6-Bromo-2,2'-bipyridyl, Sigma-Aldrich®
Catalog # SIAL690066-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.0800 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:10495-73-5
Formula:C₁₀H₇BrN₂
MDL Number:MFCD01318953