
2,6-Diaminopurine 98%
ACRO295270050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to light yellow Crystalline powder or granules
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
:
:150.14 g/mol
:302 °C
:>110 °C
:217-605-2
:GHS08
:Ambient
:1904-98-9
:MFCD00071537
:C₅H₆N₆
:MFCD00213668