0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
About
Export
ContactUsLinkComponent
Pluronic® P-123 (Synperonic® P 123)
Pluronic® P-123 (Synperonic® P123), Sigma-Aldrich®
Pluronic® P-123 (Synperonic® P123), Sigma-Aldrich®
Pluronic® P-123 (Synperonic® P123), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Pluronic® P-123 (Synperonic® P123), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
9003-11-6
:
:
M-Clarity™ quality level = MQ100
:
~5800 g/mol
Storage Temperature:
Ambient