Aprotinin
Plastic bottle for solids, 50 mg
Catalog # ROCH10981532001
Supplier: ROCHE
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 mg
- Cat. No.:ROCH10981532001
- Molecular formula:C₂₈₄H₄₃₂N₈₄O₇₉S₇
- Protein/peptide name:Aprotinin
- Storage conditions:Refrigerator
- Protein synonyms:Trypsin inhibitor, pancreatic basic
- MDL No.:MFCD00130541
- CAS No.:9087-70-1



