Order Entry
Export
ContactUsLinkComponent
Alizarin red S (sodium salt), Sigma-Aldrich®
Irritant
Alizarin red S (sodium salt), Sigma-Aldrich®
Catalog # A5533-25G
CAS Number:  
undefined
Irritant
Alizarin red S (sodium salt), Sigma-Aldrich®
Catalog # A5533-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :342.26 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:204-981-8
 :GHS07
Storage Temperature:Ambient
CAS Number:130-22-3
 :93982-72-0
Formula:C₁₄H₇NaO₇S
MDL Number:MFCD00013049