Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Catalog # ROTH7162.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g
- Seq:3122
Specifications
About this item
:
:202.35 g/mol
EINECS:200-754-2
CAS Number:71-44-3
Formula:C₁₀H₂₆N₄
MDL Number:MFCD00008215