Order Entry
Export
ContactUsLinkComponent
 
 
N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%
 
undefined
 
N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4900 to 1.4930 (20°C, 589 nm)
  • GC:
    ≥96.0 %