β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate, Sigma-Aldrich®
Catalog # SIALN1161-10VL
Supplier: Merck
CAS Number:
Specifications
- Pk:10 Vial
- Seq:0239
Specifications
About this item
:
:727.43 g/mol
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
MDL Number:MFCD03791273