Cresol red sodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:62625-29-0
:
Formula:C₂₁H₁₇NaO₅S
MDL Number:MFCD00001618
:404.42 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:263-654-8