


TRITON™ X-305 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-93-1
:
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3082
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient