Order Entry
Export
Orders LinkContactUsLinkComponent
Benzalkonium chloride

Benzalkonium chloride

Glass bottle for solids, 100 g
Catalog # ROTH2999.1
Corrosive
Irritant
Environment
Supplier:   Roth Carl
Benzalkonium chloride
Catalog # ROTH2999.1
Supplier:  Roth Carl
CAS Number:  63449-41-2
Benzalkonium chloride

Glass bottle for solids, 100 g

CAS Number:  63449-41-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
  • CAS Number:63449-41-2
  • Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
  • MDL Number:MFCD00145757
  • Melting Pt:29…34 °C
  • Density:0.98 g/cm³ (20 °C)
  • EINECS:264-151-6
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications