Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene oxide
Polyethylene oxide, M.W. 100000
Polyethylene oxide, M.W. 100000
Polyethylene oxide, M.W. 100000
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Polyethylene oxide, M.W. 100000
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
25322-68-3
:
Formula:
(-CH₂CH₂O-)n
MDL Number:
MFCD00147970
Melting Pt:
65 °C
Density:
1.4539
Storage Temperature:
Ambient
EINECS:
500-038-2
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA042236
Videos
Thermo Scientific Acroseal 25 Years
Product Literature
Thermo Scientific Cell Culture Workflow
Documentation