Adhessive wound dressings, Tegaderm™
This transparent film dressing in frame style provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses. In vitro testing shows that the transparent film provides a viral barrier from viruses 27 nm in diameter or larger while the dressing remains intact without leakage.
- Made with a hypoallergenic, latex-free adhesive that is gentle to the skin yet holds catheters and other devices in place
- Conformable: dressing flexes with skin for greater patient comfort
- Picture-frame delivery makes placement accurate and easy, minimising potential to stick to gloves or to itself