Accessories for Evolution™ UV/VIS Spectrophotometers
Supplier: Thermo Fisher Scientific
Jump to:
About this item
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- ModelForAvailabilityPrice
- SPET699-1312008-cell peltier system, Operates on all voltages and 50/60 Hz, temperature range 5...100 °C with 0.1 °C precision, 24×17×14, 35 lbs.8-cell peltier system, Operates on all voltages and 50/60 Hz, temperature range 5...100 °C with 0.1 °C precision, 24×17×14, 35 lbs.Temp/time based kinetic applicationAccessory for evolution spectrometersLog in for availability1 * 1 items11 660,00 €
- SPET222-269500Cuvette holder for ISA-220 integrating sphereCuvette holder for ISA-220 integrating sphere—Accessory for evolution 220 spectrometersLog in for availability1 * 1 items964,00 €
- SPET222-269400Evolution ISA-220 integrating sphere, Single beam optical design, 1-2 nm spectral bandwidth, 220 – 1100 nm wavelength range, 10×10 mm silicon photodiode detector, 10, 5, 2, 1,0, 0,5, 0,2, 0,1 nm data interval, 9×5×7, 1 lbs.Evolution ISA-220 integrating sphere, Single beam optical design, 1-2 nm spectral bandwidth, 220 – 1100 nm wavelength range, 10×10 mm silicon photodiode detector, 10, 5, 2, 1,0, 0,5, 0,2, 0,1 nm data interval, 9×5×7, 1 lbs.Transmission/reflectance measurementAccessory for evolution 220 spectrometersLog in for availability1 * 1 items11 240,00 €
- SPET840-326000Qualification filter kit for UV-VIS, 381×254×177 mm, 1,36 kg, 240 - 640 nm wavelength rangeQualification filter kit for UV-VIS, 381×254×177 mm, 1,36 kg, 240 - 640 nm wavelength rangeCertified standardKit to qualifify spectrophotometer for UV-Visible regionLog in for availability1 * 1 items2 350,00 €
- SPET840-341600Smart SipperSmart Sipper—Accessory for evolution one/one plus seriesLog in for availability1 * 1 items2 650,00 €
- SPET840-214800Smart thermostatted 7-cell changer, 6.1×10,8×13,8; 40 lbs.; 0...100 °C temp. range, works together with Cetac 260 and 520 autosamplers under INSIGHT Auto control for Evolution series spectrophotometersSmart thermostatted 7-cell changer, 6.1×10,8×13,8; 40 lbs.; 0...100 °C temp. range, works together with Cetac 260 and 520 autosamplers under INSIGHT Auto control for Evolution series spectrophotometersFor 7 simultaneous measurementAccessory for evolution one/one plus seriesDiscontinued-
- SPET840-219000Smart thermostatted 7-cell changer, 15×9×10, 16 lbs., 0...95 °C temp. rangeSmart thermostatted 7-cell changer, 15×9×10, 16 lbs., 0...95 °C temp. range—Accessory for evolution spectrometersLog in for availability1 * 1 items4 290,00 €
- SPET912A1199Single cell peltier unit for evolution includes Peltier cell holder, koolanceSingle cell peltier unit for evolution includes Peltier cell holder, koolanceEvolution One/One Plus/ProLog in for availability1 * 1 itemsSpecifications:New ProductNew Product