ContactUsLinkComponent
Stopcock, three-way, acc. to Czako
  WITG0.268.001
undefined
Stopcock, three-way, acc. to Czako
  WITG0.268.001
  • Hole Ø:
    1,6 mm
  • Pk:
    1
  • Spigot:
    14,5 NS
Stopcocks made of borosilicate glass.
  • With capillary side arms
  • Solid glass key
  • Good chemical and temperature resistance
 : DIN 12563