5-Fluorouridine
Glass bottle for solids, 100 mg
Catalog # MOLE22848333-100MG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione
- CAS Number:316-46-1
- Formula:C₉H₁₁FN₂O₆
- MDL Number:MFCD00036832
- Molecular Weight:262.19 g/mol
- Storage Temperature:Ambient
- EINECS:206-260-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



