Order Entry
Export
ContactUsLinkComponent
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Irritant
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # ROCH11429876001
Supplier:  ROCHE
CAS Number:  
Irritant
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # ROCH11429876001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.69 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
UN:3261
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30827-99-7
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962