3,3-Dimethylglutaric anhydride
Catalog # MOLE14541420-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:142.15 g/mol
EINECS:223-998-1
CAS Number:4160-82-1
Formula:C₇H₁₀O₃
MDL Number:MFCD00006684