Polyethylene glycol dimethylether, M.W. 250 stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:24991-55-7
:
Formula:H₃C(OCH₂CH₂)nOCH₃
MDL Number:MFCD00147972
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)