Glass fibre mats
Supplier: VOLZ LUFTFILTER
Certificates
About this item
Paint-Stop (green): For use in separation of ink mist in paint shops and colour spraying cabins. Can be used in the automotive industry, manufacturing plants for furniture, windows, machines etc. Contains no silicone or other lacquer harming substances. Fire prevention requirements according to DIN 53438 (F3).
Dust-Stop (yellow): For separation of dry dust types. Can be used in carpenters' workshops, metal working industry etc. Contains no silicone. Fire prevention requirements according to DIN 53438 (F3). Additionally impregnated with antibacterial dust adhesive.
Specifications
- Catalog No:
- VOLZMRGRUN32575
- VOLZMRGRIN425150
- VOLZMRGRIN425100
- VOLZMRGRUN32550
- VOLZMRGRUN420163
- VOLZMRGRUN320100
- VOLZPSGRUN4_125
- VOLZMRGRUN32075
- VOLZPSGRUN4110
- VOLZMRGRIN420150
- VOLZMRGRIN420100
- VOLZMRGRIN42075
- VOLZMRGRIN42050
- VOLZMRGRIN325150
- VOLZMRGRIN325100
- VOLZMRGRIN320200
- VOLZMRGRUN32050
- VOLZMRGRUN225100
- VOLZMRGRUN220150
- VOLZMRGRUN220100
- VOLZMRGRUN22075
- VOLZMRGRUN22050
- Thickness:
- 70 mm
- 100 mm
- 100 mm
- 70 mm
- 100 mm
- 70 mm
- 100 mm
- 70 mm
- 100 mm
- 100 mm
- 100 mm
- 100 mm
- 100 mm
- 70 mm
- 70 mm
- 70 mm
- 70 mm
- 50 mm
- 50 mm
- 50 mm
- 50 mm
- 50 mm
- Max. temperature:
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- 180 °C
- Average arrestance:
- 96%
- 98%
- 98%
- 96%
- 98%
- 96%
- 98%
- 96%
- 98%
- 98%
- 98%
- 98%
- 98%
- 96%
- 96%
- 96%
- 96%
- 94%
- 94%
- 94%
- 94%
- 94%