First Aid Box, Type 2
First aid kit, type 2, orange, for up to 20 people, ABS
Catalog # 118-0242
Supplier: TRENKA
Jump to:
Product Details & Documents
Printed ABS first aid kit for up to 20 workers.
- Mobile and stationary use with wall bracket and 90° stop lock
- Rubber seal: Dust and splash proof
- Transparent cover plates to protect the filling parts
- Variable interior division with dividers, sealable
- Impact and break resistant
Color: orange
H×B×T: 240×340×120 mm
: ÖNORM Z 1020 Typ 2
Specifications
- Description:First aid kit, type 2, orange, for up to 20 people
- Material:ABS
Accessories
Product Information
- Catalog #AvailabilityPrice
- Komplette Nachfüllpackung für Erste-Hilfe-Koffer Typ 2 (1 - 20 Personen)Komplette Nachfüllpackung für Erste-Hilfe-Koffer Typ 2 (1 - 20 Personen)IBSA31800016910Log in for availability1 * 1 items - 32,67 €



