Overall, Cleansafe® 1
White, XXL
Catalog # 113-08351
Supplier: TRENKA
Jump to:
Product Details & Documents
Superb™ disposable coveralls specifically designed for disposal work.
- Front zip with zip flap
- Attached socks and elasticated hood, hips and wrists
- Excellent design that finishes above the chin avoiding exposed neck areas
Größe: XXL
Schutzkleidung Typ 5+6
Certifications: Typ 5+6
Documents
Specifications
- Clothing Size:XXL
- Colour:White
- Cat. No.:113-08351
- Anti-static:+
- Garment application:Chemical protection|General Purpose|Industrial