Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Workwear
8616864
Catalog # 113-05893
Supplier:
RENTEX
Expand image
Catalog # 113-05893
Supplier:
RENTEX
Specifications
Garment application:
General Purpose
Gender:
Unisex
Colour:
Dark blue
L×W:
900×900 mm
Fabric:
Cotton
Pk:
1
Specifications
About this item
Product Details
Product Details