Knitted gloves, Nitras® 03440, fully coated, with cuff
Supplier: AS ARBEITSSCHUTZ
Certificates
About this item
Heavy work gloves made of 100% cotton jersey, fully coated with nitrile rubber.
- Offers great comfort and flexibility
- With safety cuff
- Fully coated for maximum protection
For use in the automotive industry, in manufacturing plants or in the construction trade.
EN 388 4221
Certifications: EN 388 (4221X); Cat. II
Specifications
- Catalog No:
- 112-2414
- 112-2408
- 112-2409
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton jersey
- Cotton jersey
- Cotton jersey
- Cuff style:
- Short
- Short
- Short
- EN norm:
- EN 388 (4221X)
- EN 388 (4221X)
- EN 388 (4221X)