Order Entry
Export
ContactUsLinkComponent
Desoxyribonuclease I
Desoxyribonuclease I
Catalog # ROCH11284932001
Supplier:  ROCHE
CAS Number:  
Desoxyribonuclease I
Catalog # ROCH11284932001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 mg
  • CAS No.:
    9003-98-9
  • Storage conditions:
    Freezer
  • Enzyme name:
    Desoxyribonuclease I
  • Enzyme synonyms:
    DNase I

Specifications

About this item