
Kanamycin sulphate
Catalog # MOLE15202460-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:582.6 g/mol
EINECS:246-933-9
:GHS08
Storage Temperature:Ambient
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253