Order Entry
Export
ContactUsLinkComponent
Kanamycin sulphate
Health
Kanamycin sulphate
Catalog # EHERC14505520
Supplier:  EHRENSTORFER
CAS Number:  
Health
Kanamycin sulphate
Catalog # EHERC14505520
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :582.6 g/mol
EINECS:246-933-9
 :GHS08
Storage Temperature:Ambient
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253