Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hygromycin B
Hygromycin B (from Streptomyces hygroscopicus), Sigma-Aldrich®
Hygromycin B (from Streptomyces hygroscopicus), Sigma-Aldrich®
:
Merck
Hygromycin B (from Streptomyces hygroscopicus), Sigma-Aldrich®
:
Merck
:
31282-04-9
:
:
C₂₀H₃₇N₃O₁₃
:
MFCD06795479
:
527.5 g/mol
:
6.1,I
:
M-Clarity™ quality level = MQ200
:
250-545-5
:
3462
:
GHS06
:
GHS05
:
GHS08
:
Refrigerator