Hygromycin B (from Streptomyces hygroscopicus), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:31282-04-9
:
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479
:527.5 g/mol
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:250-545-5
UN:3462
:GHS06
:GHS05
:GHS08
Storage Temperature:Refrigerator