6-Bromo-1,4-benzodioxane, Sigma-Aldrich®
Catalog # SIAL538523-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:215.05 g/mol
:M-Clarity™ quality level = MQ100
EINECS:257-817-2
CAS Number:52287-51-1
Formula:C₈H₇BrO₂
MDL Number:MFCD00040750