6-Bromo-1,4-benzodioxane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52287-51-1
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00040750
:215.05 g/mol
Boiling Pt:259…260 °C
Density:1.598
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:257-817-2