
6-Amino-1,4-benzodioxane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22013-33-8
:
Formula:C₈H₉NO₂
MDL Number:MFCD00006824
:151.17 g/mol
Boiling Pt:116…118 °C (3 mmHg)
Melting Pt:26…30 °C
Density:1.231
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:244-718-4
UN:2811
ADR:6.1,III
:GHS06