Order Entry
Export
Orders LinkContactUsLinkComponent
G418

G418

250 µl
Catalog # FITZ20-GR15
Irritant
G418
Catalog # FITZ20-GR15
CAS Number:  49863-47-0
G418

250 µl

CAS Number:  49863-47-0
Price
Inquire For Price
Log in for availability
1 * 250 µl
Product Details & Documents
Synonyms:  G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
  • CAS Number:49863-47-0
  • Formula:C₂₀H₄₀N₄O₁₀
  • MDL Number:MFCD30541267
  • Molecular Weight:496.552 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    250 µl

Specifications