Orders LinkContactUsLinkComponent
Ampoules, standard, WHEATON®
Ampoules, standard, WHEATON®
Supplier:   DWK Life Sciences
Ampoules, standard, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Ampoules, standard, WHEATON®
Ampoules, standard, WHEATON®
2 Options Available Below
Product Details & Documents
Ampoules are made from low potassium borosilicate glassare.
  • Consistent with low background counts
Commonly used for packaging certified standards.
Certifications: USP Type I, ASTM E 438 Type I, Class A
Product Family Options
Product Information
  • Colour
    Capacity
    Ø×H
    Availability
    Price
  • WHEA176759
    Ampoule, standard
    Ampoule, standard
    Clear
    7 ml
    17×100 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 620,00 €
    620,00 €
    1 * 25 items
       620,00 €
  • WHEA176762
    Ampoule, standard
    Ampoule, standard
    Clear
    20 ml
    28×100 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 857,00 €
    857,00 €
    1 * 25 items
       857,00 €