Order Entry
Export
ContactUsLinkComponent
Vitamin K1 (phytomenadione, phylloquinone)
Vitamin K1 (phytomenadione, phylloquinone)
Catalog # EHERC17924400
Supplier:  EHRENSTORFER
CAS Number:  
Vitamin K1 (phytomenadione, phylloquinone)
Catalog # EHERC17924400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :450.6958 g/mol
EINECS:201-564-2
CAS Number:84-80-0
 :81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063