Phenylmagnesium bromide ∼12% (w/w) 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-58-3
:
Formula:C₆H₅BrMg
MDL Number:MFCD00000038
:181.31 g/mol
ADR:4.3,I
:M-Clarity™ quality level = MQ200
EINECS:202-867-2
UN:3399