Order Entry
Export
ContactUsLinkComponent
 
Pesticide reference standard, λ-Cyhalothrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA11860000CY
 :  
Pesticide reference standard, λ-Cyhalothrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA11860000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    91465-08-6
  • Description:
    λ-Cyhalothrin 100 µg/ml in cyclohexane
  • Formula:
    C₂₃H₁₉ClF₃NO₃
  • Description:
    λ-Cyhalothrin