Pesticide reference standard, λ-Cyhalothrin 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL11860000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:91465-08-6
- Description:λ-Cyhalothrin 10 µg/ml in cyclohexane
- Formula:C₂₃H₁₉ClF₃NO₃
- Description:λ-Cyhalothrin