Order Entry
Export
Orders LinkContactUsLinkComponent
DL-Menthol

DL-Menthol

Glass bottle for solids, 100 g
Catalog # ROTH8391.1
Supplier:   Roth Carl
DL-Menthol
Catalog # ROTH8391.1
Supplier:  Roth Carl
CAS Number:  89-78-1
DL-Menthol

Glass bottle for solids, 100 g

CAS Number:  89-78-1
Price
37,30 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
  • CAS Number:89-78-1
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00001484
  • Molecular Weight:156.27 g/mol
  • Storage Temperature:Ambient
  • EINECS:201-939-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications