Order Entry
Export
Orders LinkContactUsLinkComponent
 

β-Cyclodextrine, Sigma-Aldrich®

Supplier:   Merck
β-Cyclodextrine, Sigma-Aldrich®
Supplier:  Merck
Certificates
β-Cyclodextrine, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:231-493-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 89,00 €
    89,00 €
    1 * 25 g
       89,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 132,00 €
    132,00 €
    1 * 100 g
       132,00 €