2-(Ethylthio)nicotinic acid
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
3 Options Available Below
Product Details & Documents
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



