Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Propylthio)nicotinoyl chloride
2-(Propylthio)nicotinoyl chloride
2-(Propylthio)nicotinoyl chloride
2-(Propylthio)nicotinoyl chloride
Supplier:
Thermo Scientific
Certificates
Expand image
2-(Propylthio)nicotinoyl chloride
Supplier:
Thermo Scientific
About this item
CAS Number:
175135-24-7
:
Formula:
C₉H₁₀ClNOS