1,3-Dichloroisoquinoline, Sigma-Aldrich®
Catalog # SIAL368830-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:198.05 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:7742-73-6
Formula:C₉H₅Cl₂N
MDL Number:MFCD00034750