Order Entry
Export
ContactUsLinkComponent
Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonate)
Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonate)
Catalog # EHERC10590000
Supplier:  EHRENSTORFER
CAS Number:  
Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonate)
Catalog # EHERC10590000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
EINECS:207-612-9
CAS Number:485-31-4
Formula:C₁₅H₁₈N₂O₆