Order Entry
Export
ContactUsLinkComponent
Bensultap
Bensultap
Catalog # EHERC10503000
Supplier:  EHRENSTORFER
CAS Number:  
Bensultap
Catalog # EHERC10503000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
CAS Number:17606-31-4
Formula:C₁₇H₂₁NO₄S₄