Herbicide reference standard, Ametryn 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10150000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:834-12-8
- Description:Ametryn 100 µg/ml in acetonitrile
- Formula:C₉H₁₇N₅S
- Description:Ametry