Order Entry
Export
ContactUsLinkComponent
1,3-Bis(trimethylsilyl)urea
1,3-Bis(trimethylsilyl)urea
Catalog # MOLEM81616581
Supplier:  Molekula
CAS Number:  
1,3-Bis(trimethylsilyl)urea
Catalog # MOLEM81616581
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :204.42 g/mol
EINECS:242-177-9
CAS Number:18297-63-7
Formula:C₇H₂₀N₂OSi₂
MDL Number:MFCD00008260