Order Entry
Export
ContactUsLinkComponent
Organic reference standard, Sulphacetamide 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, Sulphacetamide 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA16988850AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Sulphacetamide 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA16988850AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    144-80-9
  • Description:
    Sulphacetamide 100 µg/ml in acetonitrile
  • Formula:
    C₈H₁₀N₂O₃S
  • Description:
    Sulfacetamid

Specifications

About this item