Order Entry
Export
ContactUsLinkComponent
Herbicide reference standard, Chlorthal-dimethyl 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Herbicide reference standard, Chlorthal-dimethyl 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11620000CY
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Chlorthal-dimethyl 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11620000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1861-32-1
  • Description:
    Chlorthal-dimethyl 100 µg/ml in cyclohexane
  • Formula:
    C₁₀H₆C₁₄O₄
  • Description:
    Chlorthal-dimethy

Specifications

About this item