Pyrrole-3-carboxylic acid, Sigma-Aldrich®
Catalog # SIAL689416-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:111.10 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:931-03-3
Formula:C₅H₅NO₂
MDL Number:MFCD00800594